Assay Details
| Gene Symbol: Vmn2r79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105190.1 | NP_001098660.1 | 4 - 5 | 1517 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 358475 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918024 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 935451 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 658507 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 110687 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 350347 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 125500 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 180492 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 116292 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 58650 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 100076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25296413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296320 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06333984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06492458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06428640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06492460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296401 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25296346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06518463 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10554751 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10554749 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10554750 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10554748 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700010796.mm.1 | NM_001105190.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010796.mm.1 | NM_001105190.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010796.mm.1 | TC0700001250.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700010798.mm.1 | NM_001105190.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010798.mm.1 | NM_001105190.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010798.mm.1 | TC0700001250.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700010800.mm.1 | NM_001105190.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010800.mm.1 | NM_001105190.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010800.mm.1 | TC0700001250.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700010799.mm.1 | NM_001105190.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010799.mm.1 | NM_001105190.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010799.mm.1 | TC0700001250.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700010801.mm.1 | NM_001105190.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010801.mm.1 | NM_001105190.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010801.mm.1 | TC0700001250.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700010797.mm.1 | NM_001105190.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010797.mm.1 | NM_001105190.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010797.mm.1 | TC0700001250.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 749238 | - | - |

Vmn2r79
Genomic Map