Assay Details
| Gene Symbol: Pin4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027181.1 | NP_081457.1 | 1 - 2 | 104 | 149 | |
| GenBank mRNA | BC119355.1 | - | 1 - 2 | 104 | 8734104 | 149 |
| BC119357.1 | - | 1 - 2 | 104 | 8734106 | 149 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC119355.1, BC119357.1, NM_027181.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969543 | NM_027181.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969543 | AK010338.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969543 | BC119355.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969543 | BC119357.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2174258 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 594618 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 354145 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 182805 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 812368 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 652375 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136496 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2174258 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969543 | NM_027181.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969543 | AK010338.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969543 | BC119355.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969543 | BC119357.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136496 | - | - |
| Whole Mouse Genome G4122A | A_51_P270478 | NM_027181.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P270478 | AK010338.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P270478 | BC119355.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P270478 | BC119357.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32567312 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567343 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567338 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567321 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14866689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567287 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14611346 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567309 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567308 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14802623 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567312 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567368 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14891717 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14611359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567370 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567374 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567310 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14611348 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567314 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567355 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567298 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567353 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14695939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567339 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567331 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567322 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14928187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14784507 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567325 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567328 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567297 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14586182 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14552655 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14802625 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567342 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567304 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567295 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567348 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567364 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14611349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567327 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567336 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32567361 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10601273 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601274 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601276 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552285 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601275 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428658_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10601272 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10552284 | - | - |
| Mouse Genome 430 2.0 Array | 1428658_at | - | - |
| Mouse Genome 430 2.0 Array | 1428658_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00006670.mm.1 | NM_027181.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00006670.mm.1 | NM_027181.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00006670.mm.1 | TC0X00000990.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00006674.mm.1 | NM_027181.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00006674.mm.1 | NM_027181.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00006674.mm.1 | TC0X00000990.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00006673.mm.1 | NM_027181.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00006673.mm.1 | NM_027181.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00006673.mm.1 | TC0X00000990.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00006672.mm.1 | NM_027181.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00006672.mm.1 | NM_027181.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00006672.mm.1 | TC0X00000990.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428658_PM_at | - | - |
| Mouse Expression Set 430 | 1428658_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 897264 | - | - |
| ABI Mouse Genome Survey Microarray | 900376 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2691761 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2691761 | - | - |

Pin4
Genomic Map