Assay Details
| Gene Symbol: Olfr776 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207559.1 | NP_997442.1 | 1 | 456 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005615 | NM_207559.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005615 | NM_207559.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545243 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545207 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545230 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08506492 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08506485 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08506484 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08506473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545213 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08743589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08508230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08817245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545223 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10367482 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10367481 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000012025.mm.1 | NM_207559.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000012025.mm.1 | NM_207559.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000012025.mm.1 | TC1000001634.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 742195 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2901432 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2901432 | - | - |

Olfr776
Genomic Map