Assay Details
| Gene Symbol: Sec61g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109971.1 | NP_001103441.1 | 4 | 688 | 72 | |
| NM_001109972.1 | NP_001103442.1 | 4 | 667 | 72 | ||
| NM_011343.3 | NP_035473.2 | 4 | 663 | 72 | ||
| GenBank mRNA | AK012393.1 | - | 4 | 306 | 72 | |
| AK012644.1 | - | 4 | 327 | 72 | ||
| AV456975.1 | - | 4 | 350 | 72 | ||
| BC019129.1 | - | 4 | 341 | 72 | ||
| BC019158.1 | - | 4 | 341 | 3487721 | 72 | |
| BC081456.1 | - | 3 | 291 | 6407982 | 72 | |
| M88756.1 | - | 2 | 128 | 72 | ||
| U11027.1 | - | 3 | 279 | 72 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27611246 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27611245 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27611243 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27611247 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2155421 | AK012393.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2155421 | AK012644.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2155421 | AK160390.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2155421 | AV456975.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2155421 | BC019129.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 579357 | AJ006837.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 579357 | AK076188.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 579357 | BC100513.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724172 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 556918 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 16243 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147081 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2155421 | AK012393.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2155421 | AK012644.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2155421 | AK160390.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2155421 | AV456975.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2155421 | BC019129.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204390 | AK012393.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204390 | AK012644.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204390 | AV456975.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204390 | BC019129.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204390 | BC019158.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27611242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27611244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27611234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27611235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27611257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27611259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27611231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27611254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27611229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27611250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27611252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27611245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27611246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27611243 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27611240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27611247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27611232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27611248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27611225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27611261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27611237 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147081 | - | - |
| Whole Mouse Genome G4122A | A_51_P204390 | AK012393.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P204390 | AK012644.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P204390 | AV456975.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P204390 | BC019129.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P204390 | BC019158.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131881 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115384 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101185 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147081 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09046559 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09414040 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09414039 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09414034 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09414037 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09046562 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27611245 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27611246 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27611247 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27611243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27611240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09046550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09046565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09046562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09431319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09724123 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09383640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09431333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611201 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09046557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611203 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27611269 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10384449 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600719 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584279 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365715 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384450 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584278 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584277 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384451 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423090_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10584276 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10384448 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10365714 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10600718 | - | - |
| Mouse Genome 430 2.0 Array | 1423090_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1423090_x_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100021709.mm.1 | NM_001109971.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100021709.mm.1 | NM_001109971.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100021709.mm.1 | NM_001109972.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100021709.mm.1 | NM_001109972.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100021709.mm.1 | NM_011343.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100021709.mm.1 | NM_011343.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100021709.mm.1 | TC1100002240.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423090_PM_x_at | - | - |
| Mouse Expression Set 430 | 1423090_x_at | - | - |
| Murine Genome U74v2 Set | 92636_PM_f_at | - | - |
| Murine Genome U74v2 Set | 92636_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423090_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423090_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 574755 | - | - |
| ABI Mouse Genome Survey Microarray | 895528 | - | - |
| ABI Mouse Genome Survey Microarray | 903480 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4120725 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2657628 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2657628 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2657628 | - | - |

Sec61g
Genomic Map