Assay Details
| Gene Symbol: Khdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113187.1 | NP_001106658.1 | 2 - 3 | 660 | 161 | |
| GenBank mRNA | BG081161.2 | - | 2 - 3 | 386 | 161 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P196673 | NM_001113187.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P196673 | AK139842.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P196673 | NM_001113187.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P196673 | AK139842.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 811560 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 421045 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P196673 | NM_001113187.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P196673 | AK139842.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P196673 | NM_001113187.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P196673 | AK139842.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04033387 | - | - |
| Whole Mouse Genome G4122A | A_51_P196673 | NM_001113187.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P196673 | AK139842.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P196673 | NM_001113187.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P196673 | AK139842.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077733 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20077703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20077689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00430913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00317438 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20077700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20077657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20077702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00855313 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20077698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20077646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00317432 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00317431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00317448 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20077705 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00317449 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00533892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20077625 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20077715 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00748953 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00430912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00855315 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00823511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00533885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00276289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00391559 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10345081 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345080 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345086 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345085 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345084 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434163_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434164_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10345083 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10345079 | - | - |
| Mouse Genome 430 2.0 Array | 1434163_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434164_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1434163_at | - | - |
| Mouse Genome 430 2.0 Array | 1434164_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 98582_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 98582_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100000986.mm.1 | NM_001113187.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100000986.mm.1 | NM_001113187.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100000986.mm.1 | TC0100000143.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100000985.mm.1 | NM_001113187.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100000985.mm.1 | NM_001113187.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100000985.mm.1 | TC0100000143.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100000984.mm.1 | NM_001113187.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100000984.mm.1 | NM_001113187.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100000984.mm.1 | TC0100000143.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1434163_PM_at | - | - |
| Mouse Expression Set 430 | 1434164_PM_s_at | - | - |
| Mouse Expression Set 430 | 1434163_at | - | - |
| Mouse Expression Set 430 | 1434164_s_at | - | - |
| Murine Genome U74v2 Set | 94349_PM_at | - | - |
| Murine Genome U74v2 Set | 94349_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434164_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434163_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434164_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434163_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434164_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434163_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434163_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434164_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 714278 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1244800 | - | - |

Khdc1b
Genomic Map