Assay Details
| Gene Symbol: Selt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040396.2 | NP_001035486.2 | 1 - 2 | 217 | 68 | |
| GenBank mRNA | AK013022.1 | - | 1 - 2 | 181 | 68 | |
| AK032838.1 | - | 1 - 2 | 194 | 68 | ||
| AK047958.1 | - | 1 - 2 | 195 | 68 | ||
| BC019970.1 | - | 1 - 2 | 165 | 4458171 | 68 | |
| BC038867.1 | - | 1 - 2 | 93 | 1547418 | 68 | |
| BC043068.1 | - | 103 | 68 | |||
| BC132453.1 | - | 1 - 2 | 183 | 40130730 | 68 | |
| BC138424.1 | - | 1 - 2 | 183 | 8861444 | 68 | |
| CJ133950.1 | - | 1 - 2 | 218 | 68 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Selt
Genomic Map