Assay Details
| Gene Symbol: Pcgf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197992.1 | NP_932109.1 | 6 - 7 | 558 | 111 | |
| GenBank mRNA | BC028560.1 | - | 6 - 7 | 558 | 1380201 | 111 |
More Information
Updated Mapping Information
Set Membership: |
|

Pcgf1
Genomic Map