Assay Details
| Gene Symbol: Cks1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016904.1 | NP_058600.1 | 2 - 3 | 264 | 105 | |
| GenBank mRNA | AB025409.1 | - | 2 - 3 | 264 | 105 | |
| AK004101.1 | - | 2 - 3 | 250 | 105 | ||
| AK010414.2 | - | 2 - 3 | 251 | 105 | ||
| AK011314.1 | - | 2 - 3 | 253 | 105 | ||
| AK011782.1 | - | 2 - 3 | 230 | 105 | ||
| BC076579.1 | - | 2 - 3 | 256 | 6846517 | 105 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22319259 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22319260 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22319262 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2061495 | BC076579.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2061495 | AK019429.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2061495 | NM_016904.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2061495 | AB025409.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2061495 | AK004101.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P227004 | BC076579.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P227004 | AK019429.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P227004 | NM_016904.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P227004 | AB025409.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P227004 | AK004101.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P227004 | BC076579.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P227004 | AK019429.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P227004 | NM_016904.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P227004 | AB025409.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P227004 | AK004101.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 409558 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2061495 | BC076579.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2061495 | AK019429.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2061495 | NM_016904.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2061495 | AB025409.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2061495 | AK004101.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P227004 | BC076579.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P227004 | AK019429.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P227004 | NM_016904.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P227004 | AB025409.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P227004 | AK004101.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P227004 | BC076579.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P227004 | AK019429.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P227004 | NM_016904.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P227004 | AB025409.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P227004 | AK004101.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319249 | - | - |
| Whole Mouse Genome G4122A | A_51_P227004 | BC076579.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P227004 | AK019429.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P227004 | NM_016904.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P227004 | AB025409.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P227004 | AK004101.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P227004 | BC076579.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P227004 | AK019429.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P227004 | NM_016904.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P227004 | AB025409.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P227004 | AK004101.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02745710 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02745685 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02745703 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02745706 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22319262 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22319260 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22319259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319255 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319257 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319256 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02745685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03011175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319258 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319260 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02745710 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319254 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319250 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10499640 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413950 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499641 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499642 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448441_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416698_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10413949 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10499639 | - | - |
| Mouse Genome 430 2.0 Array | 1448441_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456268_at | - | - |
| Mouse Genome 430 2.0 Array | 1416698_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1448441_at | - | - |
| Mouse Genome 430 2.0 Array | 1456268_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416698_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 54124_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 54124_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300018141.mm.1 | NM_016904.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018141.mm.1 | NM_016904.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018141.mm.1 | TC0300002352.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300018140.mm.1 | NM_016904.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018140.mm.1 | NM_016904.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018140.mm.1 | TC0300002352.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300018139.mm.1 | NM_016904.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018139.mm.1 | NM_016904.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018139.mm.1 | TC0300002352.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416698_a_at | - | - |
| Mouse Expression Set 430 | 1416698_PM_a_at | - | - |
| Mouse Expression Set 430 | 1448441_PM_at | - | - |
| Mouse Expression Set 430 | 1448441_at | - | - |
| Murine Genome U74v2 Set | 97468_PM_at | - | - |
| Murine Genome U74v2 Set | 97468_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416698_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448441_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448441_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416698_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416698_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416698_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448441_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448441_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 895391 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5220300 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5220300 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106650253 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005220300 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1233989 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1233989 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225371 | - | - |
| mouse-6 v1.1 (Illumina) | scl0054124.1_261-S | - | - |
| mouse-6 v1.1 (Illumina) | scl21251.1_327-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5220300 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106650253 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0054124.1_261-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5220300 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106650253 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1233989 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1233989 | - | - |

Cks1b
Genomic Map