Assay Details
| Gene Symbol: Olfr1513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012269.2 | NP_001012269.2 | 1 | 632 | 170 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1953058 | NM_001012269.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1953058 | NM_001012269.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29606134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29606139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29606129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29606143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29606145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29606125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29606142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29606141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29606127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29606150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29606147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29606122 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11491325 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11491338 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11744377 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11732202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29606101 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606145 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606131 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606139 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606125 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606106 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11611001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606101 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606118 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11744377 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606119 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606129 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11218963 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606088 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606112 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11716275 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606114 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606117 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606110 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11716272 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606143 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11260019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11662429 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606116 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29606134 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11328928 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10419710 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10419714 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10419709 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 714787 | - | - |
| ABI Mouse Genome Survey Microarray | 887285 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3162384 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3162384 | - | - |
| mouse-6 v1.1 (Illumina) | GI_59676584-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3162384 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3162384 | - | - |

Olfr1513
Genomic Map