Assay Details
| Gene Symbol: Zgrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197997.2 | NP_932114.2 | 26 | 6831 | 145 | |
| GenBank mRNA | AK015177.1 | - | 2 | 1181 | 145 | |
| AK156938.1 | - | 11 | 2824 | 145 | ||
| BC079891.1 | - | 25 | 6943 | 145 | ||
| CO044435.1 | - | 1 | 145 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2244440 | BC082599.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2244440 | BC085187.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2244440 | BC079891.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2244440 | AY512952.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2244440 | NM_197997.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 600841 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 389452 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 556132 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 687074 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 378879 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 832920 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 847009 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 783307 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 316512 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 174391 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902613 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 888510 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 264140 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 27263 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 899244 | XM_001479285.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 899244 | XM_618996.4 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 138907 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 195803 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 390158 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 178990 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 955169 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 716751 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 550058 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 74349 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 788166 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 780315 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 323803 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 188079 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 691515 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 966137 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 565383 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 890108 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 656506 | AK018583.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 226952 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 650495 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 35243 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676646 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 40000 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 874312 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75301 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 398776 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 769953 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 48643 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 78072 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 151894 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 152389 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 93024 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 14727 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 55566 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 105114 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 157813 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 173497 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 46531 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104868 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119858 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129316 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153389 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2244440 | BC082599.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2244440 | BC085187.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2244440 | BC079891.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2244440 | AY512952.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2244440 | NM_197997.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05066116 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119858 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05066050 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00751145 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05066027 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00751242 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05066034 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129316 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104868 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153389 | - | - |
| Whole Mouse Genome G4122A | A_51_P294156 | BC082599.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P294156 | BC030185.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P294156 | BC079891.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P294156 | AK156938.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P294156 | AK046429.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P294161 | BC082599.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P294161 | BC030185.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P294161 | BC079891.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P294161 | AK156938.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P294161 | AK046429.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101157 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126265 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119858 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00751242 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104868 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129316 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509640 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509426 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509615 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509581 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509385 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509628 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02517567 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509593 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02921745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509383 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02921737 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509641 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02921762 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509620 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509408 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509376 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509589 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509642 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509406 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02760906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02543350 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03191186 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03097236 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509650 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509619 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509617 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509614 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509644 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03027111 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509621 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509423 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03161959 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02365101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22509390 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10495944 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495947 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495951 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495958 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495946 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495953 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495934 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495955 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495940 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495942 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495941 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495949 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495948 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495937 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495956 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495954 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495960 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495939 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495943 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495950 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495959 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495936 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495963 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495962 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495961 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495957 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495938 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495952 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453038_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10495945 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10495933 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10495935 | - | - |
| Mouse Genome 430 2.0 Array | 1453038_at | - | - |
| Mouse Genome 430 2.0 Array | 1453038_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71643_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71643_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300010478.mm.1 | NM_197997.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300010478.mm.1 | NM_197997.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300010478.mm.1 | TC0300001289.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300010476.mm.1 | NM_197997.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300010476.mm.1 | NM_197997.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300010476.mm.1 | TC0300001289.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300010475.mm.1 | NM_197997.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300010475.mm.1 | NM_197997.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300010475.mm.1 | TC0300001289.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300010477.mm.1 | NM_197997.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300010477.mm.1 | NM_197997.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300010477.mm.1 | TC0300001289.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1453038_PM_at | - | - |
| Mouse Expression Set 430 | 1453038_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453038_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453038_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 621368 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 103060026 | - | - |
| MouseWG-6 v2 Expression BeadChip | 102970411 | - | - |
| MouseWG-6 v2 Expression BeadChip | 104850377 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 103060026 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104850377 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104120717 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1243091 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2729860 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2523757 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1245314 | - | - |
| mouse-6 v1.1 (Illumina) | GI_37574085-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104850377 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104120717 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103060026 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104850377 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104120717 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103060026 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1245314 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1243091 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2523757 | - | - |

Zgrf1
Genomic Map