Assay Details
| Gene Symbol: Rrp7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029101.4 | NP_083377.3 | 1 | 91 | 93 | |
| GenBank mRNA | AK088456.1 | - | 1 | 70 | 93 | |
| BY280662.1 | - | 1 | 92 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK088456.1, NM_029101.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30386709 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30386713 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30386715 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30386711 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P150014 | AK036247.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P150014 | AK032596.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P150014 | AK036247.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P150014 | AK032596.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2108206 | CO042481.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952537 | AK168850.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952537 | BC012523.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952537 | AK153565.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952537 | AK159553.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952537 | AK036247.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 42514 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 572760 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 694013 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 119960 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116769 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133637 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P150014 | AK036247.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P150014 | AK032596.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P150014 | AK036247.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P150014 | AK032596.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952537 | AK168850.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952537 | BC012523.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952537 | AK153565.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952537 | AK159553.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952537 | AK036247.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2108206 | CO042481.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30386712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30386704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30386713 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30386702 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30386708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30386719 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30386715 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30386721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30386717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30386711 | - | - |
| Whole Mouse Genome G4122A | A_51_P361906 | AK168850.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P361906 | BC012523.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P361906 | AK153565.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P361906 | AK159553.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P361906 | AK036247.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P150014 | AK036247.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P150014 | AK032596.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P150014 | AK036247.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P150014 | AK032596.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137107 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12306665 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11919516 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30386711 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30386713 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30386715 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30386709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30386642 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11920877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12329375 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30386678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12320294 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30386682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30386660 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30386708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30386702 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12329385 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30386676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30386663 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30386675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30386715 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30386698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30386688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30386683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30386717 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30386692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12320310 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30386721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30386696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12329381 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30386711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30386686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30386680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12306665 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30386658 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30386706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30386685 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30386691 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11850787 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11919524 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10425792 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430944 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425797 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425791 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425796 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425790 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430942 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430943 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425793 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425798 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425794 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425795 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448141_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456814_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1444383_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10430941 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10425781 | - | - |
| Mouse Genome 430 2.0 Array | 1444383_at | - | - |
| Mouse Genome 430 2.0 Array | 1448141_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1444383_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456814_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448141_at | - | - |
| Mouse Genome 430 2.0 Array | 1456814_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74778_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74778_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500015457.mm.1 | NM_029101.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015457.mm.1 | NM_029101.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015457.mm.1 | TC1500001965.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500015469.mm.1 | NM_029101.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015469.mm.1 | NM_029101.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015469.mm.1 | TC1500001965.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500015458.mm.1 | NM_029101.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015458.mm.1 | NM_029101.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015458.mm.1 | TC1500001965.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500015474.mm.1 | NM_029101.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015474.mm.1 | NM_029101.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015474.mm.1 | TC1500001965.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500015465.mm.1 | NM_029101.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015465.mm.1 | NM_029101.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015465.mm.1 | TC1500001965.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500015461.mm.1 | NM_029101.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015461.mm.1 | NM_029101.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015461.mm.1 | TC1500001965.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500015462.mm.1 | NM_029101.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015462.mm.1 | NM_029101.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015462.mm.1 | TC1500001965.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500015459.mm.1 | NM_029101.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015459.mm.1 | NM_029101.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015459.mm.1 | TC1500001965.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500015463.mm.1 | NM_029101.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015463.mm.1 | NM_029101.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015463.mm.1 | TC1500001965.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500015466.mm.1 | NM_029101.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015466.mm.1 | NM_029101.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015466.mm.1 | TC1500001965.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500015470.mm.1 | NM_029101.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015470.mm.1 | NM_029101.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015470.mm.1 | TC1500001965.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500015472.mm.1 | NM_029101.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015472.mm.1 | NM_029101.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500015472.mm.1 | TC1500001965.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448141_PM_at | - | - |
| Mouse Expression Set 430 | 1448141_at | - | - |
| Mouse Expression Set 430 | 1456814_PM_at | - | - |
| Mouse Expression Set 430 | 1444383_PM_at | - | - |
| Mouse Expression Set 430 | 1456814_at | - | - |
| Mouse Expression Set 430 | 1444383_at | - | - |
| Murine Genome U74v2 Set | 160382_at | - | - |
| Murine Genome U74v2 Set | 160382_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448141_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448141_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448141_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448141_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456814_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456814_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444383_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444383_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1980332 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001980332 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1213124 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1213124 | - | - |
| mouse-6 v1.1 (Illumina) | scl46903.8_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1980332 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl46903.8_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1980332 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1213124 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1213124 | - | - |

Rrp7a
Genomic Map