Assay Details
| Gene Symbol: Ccr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009835.4 | NP_033965.1 | 1 - 2 | 130 | 59 | |
| GenBank mRNA | AK089712.1 | - | 1 - 2 | 131 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK089712.1, NM_009835.4 transcripts.
Set Membership: |
|

Ccr6
Genomic Map