Assay Details
| Gene Symbol: Krt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201255.2 | NP_957707.2 | 2 - 3 | 700 | 150 | |
| GenBank mRNA | AY497550.1 | - | 2 - 3 | 700 | 150 | |
More Information
Set Membership: |
|

Krt9
Genomic Map