Assay Details
| Gene Symbol: Krt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201255.2 | NP_957707.2 | 3 - 4 | 857 | 160 | |
| GenBank mRNA | AY497550.1 | - | 3 - 4 | 160 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Krt9
Genomic Map