Assay Details
| Gene Symbol: Pira6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289428.1 | NP_001276357.1 | 1068 | 135 | ||
| NM_008848.1 | NP_032874.1 | 1003 | 135 | |||
| NM_011093.1 | NP_035223.1 | 1049 | 135 | |||
| GenBank mRNA | AF041035.1 | - | 1003 | 135 | ||
| U96687.1 | - | 1049 | 135 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P18236 | U96687.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P18236 | NM_008848.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P18236 | NM_011093.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P18236 | AF041035.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P18236 | U96687.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P18236 | NM_008848.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P18236 | NM_011093.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P18236 | AF041035.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022585 | NM_001166672.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022585 | U83172.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022585 | AK170720.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022585 | AK170618.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022585 | AK161081.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022585 | NM_001166672.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022585 | U83172.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022585 | AK170720.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022585 | AK170618.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022585 | AK161081.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P18236 | U96687.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P18236 | NM_008848.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P18236 | NM_011093.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P18236 | AF041035.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P18236 | U96687.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P18236 | NM_008848.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P18236 | NM_011093.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P18236 | AF041035.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24947171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24947185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24947175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948339 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24947182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948336 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06113380 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06112773 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06112762 | - | - |
| Whole Mouse Genome G4122A | A_52_P18236 | U96687.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P18236 | NM_008848.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P18236 | NM_011093.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P18236 | AF041035.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P18236 | U96687.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P18236 | NM_008848.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P18236 | NM_011093.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P18236 | AF041035.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06112773 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06165456 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06165454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948364 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948339 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10559470 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559477 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559464 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559475 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559461 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559456 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559458 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559457 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559469 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559474 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559459 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559472 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559465 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10559467 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10559454 | - | - |
| Mouse Genome 430 2.0 Array | 1420464_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1420464_PM_s_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700000399.mm.1 | NM_008848.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000399.mm.1 | NM_008848.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000399.mm.1 | NM_011093.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000399.mm.1 | NM_011093.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000399.mm.1 | TC0700000030.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420464_PM_s_at | - | - |
| Mouse Expression Set 430 | 1420464_s_at | - | - |
| Murine Genome U74v2 Set | 100328_s_at | - | - |
| Murine Genome U74v2 Set | 100328_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420464_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420464_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 101940152 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100780537 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5670725 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4670088 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101940152 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004670088 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005670725 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2643614 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2691049 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2643614 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2691049 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1260548 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2587736 | - | - |
| mouse-6 v1.1 (Illumina) | scl0018724.1_231-S | - | - |
| mouse-6 v1.1 (Illumina) | GI_6679332-I | - | - |
| mouse-6 v1.1 (Illumina) | GI_6755067-I | - | - |
| mouse-6 v1.1 (Illumina) | scl0018729.1_229-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4670088 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100780537 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101940152 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5670725 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0018724.1_231-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0018729.1_229-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4670088 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5670725 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100780537 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101940152 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2643614 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2691049 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2587736 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2643614 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1260548 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2691049 | - | - |

Pira6
Genomic Map