Assay Details
| Gene Symbol: Zfp935 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136496.1 | NP_001129968.1 | 3 | 1509 | 84 | |
| NM_178875.4 | NP_849206.2 | 4 | 1570 | 84 | ||
| GenBank mRNA | AK018441.1 | - | 3 | 1500 | 84 | |
| AY189975.1 | - | 3 | 1486 | 84 | ||
| BC012524.1 | - | 3 | 1485 | 4008347 | 84 | |
| BC037448.1 | - | 3 | 1514 | 84 | ||
| BC047396.1 | - | 4 | 1547 | 3979342 | 84 | |
| BC087936.1 | - | 4 | 1570 | 84 | ||
| BC113193.1 | - | 4 | 1566 | 40062446 | 84 | |
| BC115966.1 | - | 1428 | 84 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185524 | BC087936.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185524 | BC113193.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185524 | BC115966.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185524 | BC012524.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185524 | BC037448.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185524 | BC087936.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185524 | BC113193.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185524 | BC115966.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185524 | BC012524.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P185524 | BC037448.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 217389 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 129758 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185524 | BC087936.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185524 | BC113193.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185524 | BC115966.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185524 | BC012524.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185524 | BC037448.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185524 | BC087936.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185524 | BC113193.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185524 | BC115966.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185524 | BC012524.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P185524 | BC037448.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576411 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576401 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09562884 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576430 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576467 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576789 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02052402 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576454 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576445 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576762 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576627 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09562862 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576986 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576308 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576458 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576506 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576668 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576647 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09562906 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09577021 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576643 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576994 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09562898 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09562877 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576744 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576495 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09577230 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576729 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576953 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576350 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576766 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576476 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576661 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576385 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09577246 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576773 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576329 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576439 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576805 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576718 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576733 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576685 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576795 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576926 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576651 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576632 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09562854 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576532 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576780 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09577004 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576546 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576425 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576502 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576676 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09562889 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576712 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576961 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576392 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09562911 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576937 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576656 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576723 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576848 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576313 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576835 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576946 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576326 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09577013 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576786 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09562870 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576484 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576638 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576470 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576664 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576913 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576552 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576843 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576966 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576918 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576519 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09562837 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576373 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576441 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576537 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576859 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09576900 | - | - |
| Whole Mouse Genome G4122A | A_51_P185524 | BC087936.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P185524 | BC113193.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P185524 | BC115966.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P185524 | BC012524.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P185524 | BC037448.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P185524 | BC087936.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P185524 | BC113193.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P185524 | BC115966.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P185524 | BC012524.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P185524 | BC037448.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P306446 | AK033958.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P306446 | BC053725.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P306446 | NM_173381.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P306446 | AK033001.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P672351 | BC087936.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P672351 | BC115966.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P672351 | BC012524.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P672351 | BC037448.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P672351 | BC047396.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104264 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104832 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09577230 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576780 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576744 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576859 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576682 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576766 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09562889 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576455 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576496 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576425 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576430 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576386 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576718 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576308 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576347 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576795 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576481 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576537 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576729 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576373 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09562870 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576401 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576627 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576835 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576676 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576329 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09577019 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576805 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576986 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576519 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576946 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02052402 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576913 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09562911 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09576552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P33007349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P33007343 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10409974 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409971 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409977 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10405736 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409981 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409983 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409972 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409973 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409975 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409976 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430074_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10405733 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10409970 | - | - |
| Mouse Genome 430 2.0 Array | 1430074_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1430074_PM_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71508_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71508_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300013896.mm.1 | NM_001136496.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013896.mm.1 | NM_001136496.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013896.mm.1 | NM_178875.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013896.mm.1 | NM_178875.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013896.mm.1 | TC1300002767.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300013900.mm.1 | NM_001136496.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013900.mm.1 | NM_001136496.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013900.mm.1 | NM_178875.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013900.mm.1 | NM_178875.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013900.mm.1 | TC1300002767.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300013897.mm.1 | NM_178875.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013897.mm.1 | NM_178875.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013897.mm.1 | TC1300002767.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300013898.mm.1 | NM_001136496.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013898.mm.1 | NM_001136496.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013898.mm.1 | NM_178875.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013898.mm.1 | NM_178875.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013898.mm.1 | TC1300002767.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300013894.mm.1 | NM_001136496.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013894.mm.1 | NM_001136496.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013894.mm.1 | NM_178875.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013894.mm.1 | NM_178875.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013894.mm.1 | TC1300002767.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300013895.mm.1 | NM_001136496.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013895.mm.1 | NM_001136496.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013895.mm.1 | NM_178875.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013895.mm.1 | NM_178875.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013895.mm.1 | TC1300002767.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300013893.mm.1 | NM_001136496.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013893.mm.1 | NM_001136496.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013893.mm.1 | NM_178875.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013893.mm.1 | NM_178875.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300013893.mm.1 | TC1300002767.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1430074_PM_x_at | - | - |
| Mouse Expression Set 430 | 1430074_x_at | - | - |
| Murine Genome U74v2 Set | 141163_r_at | - | - |
| Murine Genome U74v2 Set | 141162_PM_f_at | - | - |
| Murine Genome U74v2 Set | 141162_f_at | - | - |
| Murine Genome U74v2 Set | 141163_PM_r_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430074_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430074_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 388184 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2693167 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2542129 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2693167 | - | - |

Zfp935
Genomic Map