| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700036326.mm.1 |
NM_198011.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036326.mm.1 |
NM_198011.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036326.mm.1 |
TC0700004663.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700039199.mm.1 |
NM_198011.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700039199.mm.1 |
NM_198011.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700039199.mm.1 |
TC0700004663.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700036320.mm.1 |
NM_198011.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036320.mm.1 |
NM_198011.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036320.mm.1 |
TC0700004663.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700036327.mm.1 |
NM_198011.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036327.mm.1 |
NM_198011.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036327.mm.1 |
TC0700004663.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700036323.mm.1 |
NM_198011.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036323.mm.1 |
NM_198011.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036323.mm.1 |
TC0700004663.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700036324.mm.1 |
NM_198011.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036324.mm.1 |
NM_198011.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036324.mm.1 |
TC0700004663.mm.1 |
MTA TC |