Assay Details
| Gene Symbol: Lrrc38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162983.1 | NP_001156455.1 | 1 - 2 | 1062 | 83 | |
| GenBank mRNA | AK038658.1 | - | 1 - 2 | 1026 | 83 | |
| AK045936.1 | - | 1 - 2 | 614 | 83 | ||
| AK078190.1 | - | 1 - 2 | 1037 | 83 | ||
| AK160116.1 | - | 1 - 2 | 152 | 83 | ||
| BC129963.1 | - | 1 - 2 | 1055 | 83 | ||
| BC129964.1 | - | 2 - 3 | 985 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P446106 | AK160116.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P446106 | AK038658.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P446106 | AK045936.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P446106 | AK078190.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P446106 | NM_001162983.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P446106 | AK160116.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P446106 | AK038658.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P446106 | AK045936.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P446106 | AK078190.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P446106 | NM_001162983.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 310694 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 541930 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 595191 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 702891 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 700281 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 536337 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 14996 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 157664 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 8047 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 817284 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 795554 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 798306 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 667383 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 273432 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 91897 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152709 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150166 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P446106 | AK160116.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P446106 | AK038658.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P446106 | AK045936.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P446106 | AK078190.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P446106 | NM_001162983.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P446106 | AK160116.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P446106 | AK038658.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P446106 | AK045936.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P446106 | AK078190.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P446106 | NM_001162983.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152709 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150166 | - | - |
| Whole Mouse Genome G4122A | A_51_P446106 | AK160116.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P446106 | AK038658.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P446106 | AK045936.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P446106 | AK078190.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P446106 | NM_001162983.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P446106 | AK160116.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P446106 | AK038658.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P446106 | AK045936.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P446106 | AK078190.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P446106 | NM_001162983.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150166 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04057796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04033400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03313263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04067574 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374677 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03620853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374808 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374715 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374734 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03367791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03860215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04080737 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03779507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03641756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03968201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04057800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374742 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03710564 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374721 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374806 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374743 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03641757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04080736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374733 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03851246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374713 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03952907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03505380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03968198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03779505 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03644242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03505387 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04000858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04057797 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03313258 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03860214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03505384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03858152 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374753 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374739 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03779513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04076752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03779503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03620869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03968202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03620871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374748 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03860212 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04067575 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374809 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374732 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03620866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03658186 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03620862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03505382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374683 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03461968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03753874 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374738 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03779509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04067578 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374744 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03710562 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374704 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23374701 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10510055 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10510054 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439491_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10510053 | - | - |
| Mouse Genome 430 2.0 Array | 1439491_at | - | - |
| Mouse Genome 430 2.0 Array | 1439491_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 242735_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 242735_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400015043.mm.1 | NM_001162983.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400015043.mm.1 | NM_001162983.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400015043.mm.1 | TC0400001822.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400015044.mm.1 | NM_001162983.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400015044.mm.1 | NM_001162983.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400015044.mm.1 | TC0400001822.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1439491_at | - | - |
| Mouse Expression Set 430 | 1439491_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439491_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439491_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 584251 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2648524 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2648524 | - | - |

Lrrc38
Genomic Map