Assay Details
Gene Symbol: Gpr55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK032227.1 | - | 1 - 2 | 2882 | 90 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4933407L21 gene | 4933407L21Rik | -1.5kb | Anti-sense | Chr. 1 - 85928483 - 85931756 on NCBI Build GRCm38 | |
Downstream | spermatogenesis associated 3 | Spata3 |
1700011N12Rik, 1700029H01Rik, 4930424D10Rik, Mtsarg1, Tsarg1 |
80.5kb | Anti-sense | Chr. 1 - 86017840 - 86029958 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
