Assay Details
| Gene Symbol: Chst9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199055.2 | NP_951010.1 | 2 - 3 | 181 | 128 | |
| GenBank mRNA | AB106879.1 | - | 2 - 3 | 181 | 128 | |
| BC107361.1 | - | 2 - 3 | 179 | 40056052 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chst9
Genomic Map