Assay Details
| Gene Symbol: Ube2j2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284312.1 | NP_001271241.1 | 1 - 2 | 104 | 60 | |
| GenBank mRNA | BC065779.1 | - | 1 - 2 | 104 | 1429290 | 60 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC065779.1, NM_001284312.1 transcripts.
Set Membership: |
|

Ube2j2
Genomic Map