Assay Details
| Gene Symbol: Fignl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021891.3 | NP_068691.2 | 1 - 2 | 66 | 77 | |
| GenBank mRNA | AK051324.1 | - | 1 - 2 | 37 | 77 | |
| AK051874.1 | - | 1 - 2 | 36 | 77 | ||
| AK086769.1 | - | 1 - 2 | 64 | 77 | ||
| AK088574.1 | - | 1 - 2 | 49 | 77 | ||
| CN693395.1 | - | 1 - 2 | 189 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK051874.1, AK088574.1, NM_021891.3 transcripts.
Set Membership: |
|

Fignl1
Genomic Map