Assay Details
| Gene Symbol: Ins1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008386.3 | NP_032412.3 | 2 | 533 | 80 | |
| GenBank mRNA | AK007482.1 | - | 2 | 80 | ||
| BP773571.1 | - | 1 | 80 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1959318 | BP773571.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1959318 | BC098468.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1959318 | BC145868.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1959318 | BC145870.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1959318 | S70393.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740997 | BC111097.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740997 | BC127233.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740997 | BC127234.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740997 | NM_146834.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740997 | NM_207673.1 | refseq_mrna_mouse_NM |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158374 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1959318 | BP773571.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1959318 | BC098468.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1959318 | BC145868.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1959318 | BC145870.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1959318 | S70393.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P312755 | BP773571.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P312755 | BC098468.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P312755 | BC145868.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P312755 | BC145870.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P312755 | NM_008386.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176217 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32176251 | - | - |
| Whole Mouse Genome G4122A | A_51_P312755 | BP773571.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P312755 | BC098468.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P312755 | BC145868.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P312755 | BC145870.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P312755 | NM_008386.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117200 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129691 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14382196 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14382218 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14382219 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14382221 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14382201 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14382199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32176240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176298 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176263 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176267 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176211 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176271 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14416079 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176251 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176265 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176277 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176209 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176285 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176204 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176203 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176234 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176269 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14382204 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176279 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176233 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14428766 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176290 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176273 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176225 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176287 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176246 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176231 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176295 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176214 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176223 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176293 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176227 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176216 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176205 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176218 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176255 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176236 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176275 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176281 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176202 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176215 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32176244 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10463908 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10463910 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10463909 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10463907 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422447_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10463906 | - | - |
| Mouse Genome 430 2.0 Array | 1422447_at | - | - |
| Mouse Genome 430 2.0 Array | 1422447_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16333_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16333_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900006582.mm.1 | NM_008386.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900006582.mm.1 | TC1900000743.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422447_at | - | - |
| Mouse Expression Set 430 | 1422447_PM_at | - | - |
| Murine Genome U74v2 Set | 97659_PM_r_at | - | - |
| Murine Genome U74v2 Set | 97658_f_at | - | - |
| Murine Genome U74v2 Set | 97658_PM_f_at | - | - |
| Murine Genome U74v2 Set | 97659_r_at | - | - |
| Murine Genome U74v2 Set | 165489_PM_f_at | - | - |
| Murine Genome U74v2 Set | 165489_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422447_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422447_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422447_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422447_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2060148 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002060148 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2743236 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2743236 | - | - |
| mouse-6 v1.1 (Illumina) | scl52937.2.1_149-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2060148 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl52937.2.1_149-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2060148 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2743236 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2743236 | - | - |

Ins1
Genomic Map