Assay Details
| Gene Symbol: Mrgprb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175531.4 | NP_780740.2 | 2 | 1841 | 137 | |
| GenBank mRNA | AK029369.1 | - | 2 | 1842 | 137 | |
| BX519265.1 | - | 1 | 115 | 137 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P389650 | AK029369.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P389650 | BC107390.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P389650 | BC107391.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P389650 | NM_175531.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P389650 | AK029369.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P389650 | BC107390.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P389650 | BC107391.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P389650 | NM_175531.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 96692 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630691 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 685364 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 514015 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 65216 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168161 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P389650 | AK029369.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P389650 | BC107390.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P389650 | BC107391.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P389650 | NM_175531.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P389650 | AK029369.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P389650 | BC107390.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P389650 | BC107391.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P389650 | NM_175531.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P389650 | AK029369.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P389650 | BC107390.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P389650 | BC107391.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P389650 | NM_175531.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P389650 | AK029369.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P389650 | BC107390.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P389650 | BC107391.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P389650 | NM_175531.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25107517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25107494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25107513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25107510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25107511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25107508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25107497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25107504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25107514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25107519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25107518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25107493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25107499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25107501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25107500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25107520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25107502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25107492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25107505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25107507 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25107516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25107512 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25107521 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06189717 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06189702 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168161 | - | - |
| Whole Mouse Genome G4122A | A_51_P389650 | AK029369.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P389650 | BC107390.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P389650 | BC107391.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P389650 | NM_175531.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P389650 | AK029369.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P389650 | BC107390.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P389650 | BC107391.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P389650 | NM_175531.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126090 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168161 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06356968 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06356971 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06356967 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06356954 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06356960 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06356973 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06356969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25107501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107512 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06260045 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107480 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06260037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25107521 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10563761 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563762 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563763 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10563760 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700028014.mm.1 | NM_175531.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700028014.mm.1 | NM_175531.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700028014.mm.1 | TC0700003070.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700028013.mm.1 | NM_175531.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700028013.mm.1 | NM_175531.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700028013.mm.1 | TC0700003070.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700028015.mm.1 | NM_175531.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700028015.mm.1 | NM_175531.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700028015.mm.1 | TC0700003070.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 167910_at | - | - |
| Murine Genome U74v2 Set | 167910_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 759865 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4060095 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004060095 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1257975 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1257975 | - | - |
| mouse-6 v1.1 (Illumina) | scl31324.2.1_106-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4060095 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31324.2.1_106-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4060095 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1257975 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1257975 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl31324.2.1_106-S | - | - |

Mrgprb2
Genomic Map