Assay Details
| Gene Symbol: Sprr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009264.2 | NP_033290.1 | 2 | 720 | 94 | |
| GenBank mRNA | AK144782.1 | - | 2 | 734 | 94 | |
| BC027534.1 | - | 2 | 720 | 1362548 | 94 | |
| X91824.1 | - | 2 | 713 | 94 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139678 | X91824.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139678 | NM_009264.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139678 | AK144782.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139678 | BC027534.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139678 | X91824.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139678 | NM_009264.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139678 | AK144782.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139678 | BC027534.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 496098 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75032 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115003 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139678 | X91824.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139678 | NM_009264.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139678 | AK144782.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139678 | BC027534.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139678 | X91824.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139678 | NM_009264.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139678 | AK144782.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139678 | BC027534.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331640 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331645 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331654 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331643 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331655 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331686 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331657 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22331675 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115003 | - | - |
| Whole Mouse Genome G4122A | A_51_P139678 | X91824.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139678 | NM_009264.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139678 | AK144782.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139678 | BC027534.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139678 | X91824.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139678 | NM_009264.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139678 | AK144782.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139678 | BC027534.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118927 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115003 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02850728 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02850749 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02850741 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02850746 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02850745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22331590 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331691 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02850756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331645 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331609 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331622 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331654 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331619 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331668 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331639 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331713 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331640 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331614 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02643466 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331618 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331601 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03013721 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331711 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03008423 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02846090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331628 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331637 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02687949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02850745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331683 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331669 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331634 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331643 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02438919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02786063 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331710 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331616 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331612 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331605 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331625 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331600 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02850736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22331692 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10499900 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499901 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449133_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10499899 | - | - |
| Mouse Genome 430 2.0 Array | 1449133_at | - | - |
| Mouse Genome 430 2.0 Array | 1449133_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20753_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20753_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300018570.mm.1 | NM_009264.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018570.mm.1 | NM_009264.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018570.mm.1 | TC0300002393.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300018571.mm.1 | NM_009264.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018571.mm.1 | NM_009264.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018571.mm.1 | TC0300002393.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449133_PM_at | - | - |
| Mouse Expression Set 430 | 1449133_at | - | - |
| Murine Genome U74v2 Set | 160909_PM_at | - | - |
| Murine Genome U74v2 Set | 160909_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449133_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449133_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449133_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449133_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 824922 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3870064 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003870064 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225890 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225890 | - | - |
| mouse-6 v1.1 (Illumina) | scl21900.2.1_11-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3870064 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl21900.2.1_11-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3870064 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2975250 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2975250 | - | - |

Sprr1a
Genomic Map