| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100034889.mm.1 |
NM_010410.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034889.mm.1 |
NM_010410.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034889.mm.1 |
TC1100003784.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100034887.mm.1 |
NM_010410.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034887.mm.1 |
NM_010410.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034887.mm.1 |
TC1100003784.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100034888.mm.1 |
NM_010410.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034888.mm.1 |
NM_010410.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034888.mm.1 |
TC1100003784.mm.1 |
MTA TC |