| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700029458.mm.1 |
NR_040340.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700029458.mm.1 |
NR_040340.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700029458.mm.1 |
NR_040341.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700029458.mm.1 |
NR_040341.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700029458.mm.1 |
TC0700003459.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700029461.mm.1 |
NR_040341.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700029461.mm.1 |
NR_040341.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700029461.mm.1 |
TC0700003459.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700029457.mm.1 |
NR_040340.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700029457.mm.1 |
NR_040340.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700029457.mm.1 |
NR_040341.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700029457.mm.1 |
NR_040341.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700029457.mm.1 |
TC0700003459.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700029456.mm.1 |
NR_040340.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700029456.mm.1 |
NR_040340.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700029456.mm.1 |
NR_040341.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700029456.mm.1 |
NR_040341.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700029456.mm.1 |
TC0700003459.mm.1 |
MTA TC |