Assay Details
| Gene Symbol: Tmsb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039392.2 | NP_001034481.1 | 1 | 306 | 117 | |
| NM_001190327.1 | NP_001177256.1 | 3 | 317 | 117 | ||
| NM_025284.4 | NP_079560.1 | 2 | 245 | 117 | ||
| GenBank mRNA | AK003178.1 | - | 2 | 162 | 117 | |
| AK005512.1 | - | 2 | 162 | 117 | ||
| AK008557.1 | - | 2 | 162 | 117 | ||
| AK012361.1 | - | 2 | 162 | 117 | ||
| AK019212.1 | - | 2 | 162 | 117 | ||
| AK019232.1 | - | 2 | 163 | 117 | ||
| AK131711.1 | - | 2 | 162 | 117 | ||
| AK161987.1 | - | 2 | 162 | 117 | ||
| BC037153.1 | - | 2 | 153 | 117 | ||
| BC038926.1 | - | 2 | 163 | 117 | ||
| BC048070.1 | - | 2 | 141 | 117 | ||
| BC059777.1 | - | 2 | 149 | 117 | ||
| BC070416.1 | - | 2 | 140 | 117 | ||
| BC080312.1 | - | 2 | 143 | 117 | ||
| BC089326.1 | - | 2 | 121 | 117 | ||
| BC089327.1 | - | 1 | 101 | 117 | ||
| BC093587.1 | - | 2 | 147 | 117 | ||
| BC094284.1 | - | 2 | 135 | 117 | ||
| BC099374.1 | - | 2 | 144 | 117 | ||
| BC145877.1 | - | 2 | 122 | 40131180 | 117 | |
| BC145879.1 | - | 2 | 122 | 40131182 | 117 | |
| CA466695.1 | - | 309 | 117 | |||
| CB273987.1 | - | 1 | 252 | 117 | ||
| CJ042420.1 | - | 1 | 248 | 117 | ||
| Z48496.1 | - | 1 | 239 | 116 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24544424 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24544423 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24544422 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24544425 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24544421 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24544426 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24544419 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2092085 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2092085 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2168990 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P163489 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419711 | CA466695.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419711 | CB273987.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419711 | BC092009.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419711 | BC093587.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419711 | BC094284.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419711 | CA466695.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419711 | CB273987.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419711 | BC092009.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419711 | BC093587.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419711 | BC094284.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P163489 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24544425 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24544426 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24544424 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24544422 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24544419 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24544423 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24544421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24544397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05791737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05509944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05435595 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05762141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544380 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05786506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05424746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05108646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05311730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05212783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05424744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544382 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05182397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544410 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05527256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05108651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05741198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24544449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05807042 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10545480 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545482 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545481 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551010 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545483 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417219_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10551009 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10545479 | - | - |
| Mouse Genome 430 2.0 Array | 1436902_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1455946_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1437185_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1455946_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1436902_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1417219_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1417219_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1436682_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1436682_at | - | - |
| Mouse Genome 430 2.0 Array | 1437185_s_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600019425.mm.1 | NM_001039392.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019425.mm.1 | NM_001039392.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019425.mm.1 | NM_001190327.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019425.mm.1 | NM_001190327.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019425.mm.1 | NM_025284.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019425.mm.1 | NM_025284.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019425.mm.1 | TC0600002504.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600019428.mm.1 | NM_001039392.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019428.mm.1 | NM_001039392.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019428.mm.1 | TC0600002504.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600019424.mm.1 | NM_001039392.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019424.mm.1 | NM_001039392.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019424.mm.1 | NM_001190327.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019424.mm.1 | NM_001190327.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019424.mm.1 | NM_025284.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019424.mm.1 | NM_025284.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019424.mm.1 | TC0600002504.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600019427.mm.1 | NM_001039392.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019427.mm.1 | NM_001039392.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019427.mm.1 | NM_001190327.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019427.mm.1 | NM_001190327.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019427.mm.1 | NM_025284.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019427.mm.1 | NM_025284.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019427.mm.1 | TC0600002504.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600019435.mm.1 | NM_001190327.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019435.mm.1 | NM_001190327.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019435.mm.1 | TC0600002504.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600019433.mm.1 | NM_025284.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019433.mm.1 | NM_025284.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019433.mm.1 | TC0600002504.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600019431.mm.1 | NM_001190327.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019431.mm.1 | NM_001190327.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019431.mm.1 | NM_025284.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019431.mm.1 | NM_025284.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600019431.mm.1 | TC0600002504.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417219_PM_s_at | - | - |
| Mouse Expression Set 430 | 1455946_x_at | - | - |
| Mouse Expression Set 430 | 1436902_x_at | - | - |
| Mouse Expression Set 430 | 1455946_PM_x_at | - | - |
| Mouse Expression Set 430 | 1436682_PM_at | - | - |
| Mouse Expression Set 430 | 1417219_s_at | - | - |
| Mouse Expression Set 430 | 1436682_at | - | - |
| Mouse Expression Set 430 | 1437185_s_at | - | - |
| Mouse Expression Set 430 | 1437185_PM_s_at | - | - |
| Mouse Expression Set 430 | 1436902_PM_x_at | - | - |
| Murine Genome U74v2 Set | 98129_PM_at | - | - |
| Murine Genome U74v2 Set | 98129_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437185_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436682_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436902_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436682_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455946_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417219_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436902_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455946_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417219_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437185_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 333616 | - | - |
| ABI Mouse Genome Survey Microarray | 897376 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 101780041 | - | - |
| MouseWG-6 v2 Expression BeadChip | 2340484 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101780041 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2340484 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002340484 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2476238 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225332 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2476238 | - | - |
| mouse-6 v1.1 (Illumina) | scl0019240.2_329-S | - | - |
| mouse-6 v1.1 (Illumina) | scl28849.4_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101780041 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2340484 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0019240.2_329-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101780041 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2340484 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1225332 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3145331 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3067068 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225332 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3145331 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3067068 | - | - |

Tmsb10
Genomic Map