Assay Details
| Gene Symbol: Olfr952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146503.1 | NP_666714.1 | 1 | 67 | 125 | |
| GenBank mRNA | BC150723.1 | - | 1 | 145 | 9087634 | 125 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Olfr952
Genomic Map