Assay Details
| Gene Symbol: Wnk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175638.3 | NP_783569.1 | 3 - 4 | 1110 | 80 | |
| GenBank mRNA | AY184228.1 | - | 3 - 4 | 1079 | 80 | |
| AY187027.1 | - | 3 - 4 | 1035 | 80 | ||
| AY260059.1 | - | 3 - 4 | 1073 | 80 | ||
| BC037114.1 | - | 3 - 4 | 1070 | 80 | ||
| BC043677.1 | - | 3 - 4 | 483 | 80 | ||
| BC096453.1 | - | 3 - 4 | 1066 | 4234311 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wnk4
Genomic Map