Assay Details
| Gene Symbol: Wnk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175638.3 | NP_783569.1 | 8 - 9 | 1959 | 60 | |
| GenBank mRNA | AY184228.1 | - | 8 - 9 | 1928 | 60 | |
| AY187027.1 | - | 8 - 9 | 1884 | 60 | ||
| AY260059.1 | - | 8 - 9 | 1922 | 60 | ||
| BC026679.1 | - | 1 - 2 | 158 | 60 | ||
| BC037114.1 | - | 8 - 9 | 1919 | 60 | ||
| BC043677.1 | - | 8 - 9 | 1332 | 60 | ||
| BC096453.1 | - | 8 - 9 | 1915 | 4234311 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wnk4
Genomic Map