Assay Details
| Gene Symbol: Mrgprx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207540.3 | NP_997423.1 | 2 | 435 | 74 | |
| GenBank mRNA | AY152435.1 | - | 2 | 441 | 74 | |
| BC150669.1 | - | 2 | 382 | 9087580 | 74 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991555 | NM_207540.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991555 | AY152435.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 873027 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 322442 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 23229 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991555 | NM_207540.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991555 | AY152435.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25106192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25106203 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25106185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25106165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25106189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25106200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25106194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25106168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25106202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25106182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25106190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25106181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25106195 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25106173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25106177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25106198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25106204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25106175 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06189098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25106194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06370072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06526910 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106203 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06168754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05919551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06009454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06526913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06553792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06553788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05919553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05990377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05919548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25106168 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10563742 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563741 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10563740 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700027980.mm.1 | NM_207540.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700027980.mm.1 | NM_207540.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700027980.mm.1 | TC0700003061.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700027982.mm.1 | NM_207540.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700027982.mm.1 | NM_207540.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700027982.mm.1 | TC0700003061.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700027981.mm.1 | NM_207540.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700027981.mm.1 | NM_207540.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700027981.mm.1 | TC0700003061.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 105900601 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1233345 | - | - |
| mouse-6 v1.1 (Illumina) | GI_46430535-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105900601 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105900601 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2941546 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1233345 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2941546 | - | - |

Mrgprx1
Genomic Map