Assay Details
| Gene Symbol: Odam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027128.2 | NP_081404.2 | 6 - 7 | 533 | 65 | |
| GenBank mRNA | AK009298.1 | - | 3 - 4 | 237 | 65 | |
| BC132394.1 | - | 3 - 4 | 177 | 40130671 | 65 | |
| BC132396.1 | - | 3 - 4 | 176 | 40130673 | 65 | |
| EF113909.1 | - | 6 - 7 | 533 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Odam
Genomic Map