Assay Details
| Gene Symbol: Odam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027128.2 | NP_081404.2 | 7 - 8 | 583 | 76 | |
| GenBank mRNA | AK009298.1 | - | 4 - 5 | 287 | 76 | |
| BC132394.1 | - | 4 - 5 | 227 | 40130671 | 76 | |
| BC132396.1 | - | 4 - 5 | 226 | 40130673 | 76 | |
| EF113909.1 | - | 7 - 8 | 583 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Odam
Genomic Map