Assay Details
| Gene Symbol: Odam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK009298.1 | - | 7 - 8 | 576 | 107 | |
| BC132396.1 | - | 7 - 8 | 515 | 40130673 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Odam
Genomic Map