Assay Details
| Gene Symbol: Ldlrad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033979.1 | NP_001029151.1 | 3 - 4 | 386 | 79 | |
| GenBank mRNA | AK132881.1 | - | 3 - 4 | 386 | 79 | |
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23340684 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23340687 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23340686 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055227 | AK132881.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055227 | NM_001033979.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 844454 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 153438 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 62804 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055227 | AK132881.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055227 | NM_001033979.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05420102 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05420102 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23340686 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23340684 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23340687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23340653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23340677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23340662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23340686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23340663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23340705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23340692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23340684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23340648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23340699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23340650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23340703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23340675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23340700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23340681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23340702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23340698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23340706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23340666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23340657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23340707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23340682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23340689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340704 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03607470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340656 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340690 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03873959 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03325751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03607477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03369719 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340705 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03607478 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03840746 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03781270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340669 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340677 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03607472 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03873960 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340697 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340671 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03544298 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340691 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03369717 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340692 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03369722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04025713 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04094584 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03806185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23340682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04025714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03544299 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10517580 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517582 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517581 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517583 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10517579 | - | - |
| Mouse Genome 430 2.0 Array | 1445832_at | - | - |
| Mouse Genome 430 2.0 Array | 1445832_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2922672 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2922672 | - | - |

Ldlrad2
Genomic Map