Assay Details
| Gene Symbol: Mex3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039214.4 | NP_001034303.3 | 1 | 1049 | 49 | |
| GenBank mRNA | AK133686.1 | - | 1 | 966 | 49 | |
| BC035207.1 | - | 1 | 164 | 49 | ||
| BC125427.1 | - | 1 | 331 | 40129942 | 49 | |
| KC456334.1 | - | 2 | 110 | 49 | ||
| KC456335.1 | - | 2 | 90 | 49 | ||
| KC456336.1 | - | 2 | 200 | 49 | ||
| KC456337.1 | - | 2 | 200 | 49 | ||
| KC456338.1 | - | 2 | 134 | 49 | ||
| KC456339.1 | - | 2 | 197 | 49 | ||
| KC456340.1 | - | 2 | 78 | 49 | ||
| KC456341.1 | - | 2 | 159 | 49 | ||
| KC456342.1 | - | 1 | 296 | 49 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the KC456334.1, KC456335.1, KC456336.1, KC456337.1, KC456338.1, KC456340.1, KC456341.1 transcripts.
Set Membership: |
|

Mex3c
Genomic Map