Assay Details
| Gene Symbol: Rpl12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009076.3 | NP_033102.2 | 3 - 4 | 298 | 83 | |
| GenBank mRNA | AK002973.1 | - | 3 - 4 | 297 | 83 | |
| AK008347.1 | - | 3 - 4 | 297 | 83 | ||
| AK012349.1 | - | 3 - 4 | 297 | 83 | ||
| AK012428.1 | - | 3 - 4 | 295 | 83 | ||
| AK088464.1 | - | 3 - 4 | 297 | 83 | ||
| AK144905.1 | - | 3 - 4 | 297 | 83 | ||
| AK148566.1 | - | 499 | 83 | |||
| AK150318.1 | - | 3 - 4 | 298 | 83 | ||
| AK151532.1 | - | 3 - 4 | 297 | 83 | ||
| AK151592.1 | - | 3 - 4 | 297 | 83 | ||
| AK167761.1 | - | 3 - 4 | 317 | 83 | ||
| AK168121.1 | - | 3 - 4 | 297 | 83 | ||
| AK194205.1 | - | 1 - 2 | 58 | 83 | ||
| BC018321.1 | - | 3 - 4 | 294 | 3583073 | 83 | |
| BC075731.1 | - | 2 - 3 | 486 | 83 | ||
| BC081469.1 | - | 3 - 4 | 281 | 6824411 | 83 | |
| BC090393.1 | - | 276 | 5011867 | 83 | ||
| L04280.1 | - | 3 - 4 | 285 | 83 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl12
Genomic Map