Assay Details
| Gene Symbol: Gpr55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033290.2 | NP_001028462.2 | 2 | 781 | 102 | |
| GenBank mRNA | AK146484.1 | - | 2 | 734 | 102 | |
| BC115578.1 | - | 2 | 621 | 40102229 | 102 | |
| BC115579.1 | - | 2 | 621 | 40102231 | 102 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
G-protein coupled receptor signaling pathway
activation of phospholipase C activity
positive regulation of Rho protein signal transduction
cannabinoid signaling pathway
bone resorption
negative regulation of osteoclast differentiation
positive regulation of ERK1 and ERK2 cascade
signal transducer activity
G-protein coupled receptor activity
cannabinoid receptor activity
signal transduction
G-protein coupled receptor signaling pathway
activation of phospholipase C activity
positive regulation of Rho protein signal transduction
cannabinoid signaling pathway
bone resorption
negative regulation of osteoclast differentiation
positive regulation of ERK1 and ERK2 cascade
signal transducer activity
G-protein coupled receptor activity
cannabinoid receptor activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087850 | BY759107.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087850 | NM_001033290.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087850 | AK146484.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 400162 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 272027 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 573952 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 871841 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 378666 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 73952 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 652738 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 342797 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 581158 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 636092 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 402901 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 773992 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 649216 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 788849 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 604069 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 93444 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 34197 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 57855 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087850 | BY759107.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087850 | NM_001033290.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087850 | AK146484.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00122493 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04173650 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00122503 | - | - |
| Whole Mouse Genome G4122A | A_52_P382067 | BC115579.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P382067 | NM_001033290.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P382067 | AK146484.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04173650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20409498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00308464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409390 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409490 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00901247 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409307 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409447 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00424889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409452 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00212518 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00026090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00175217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409271 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00212521 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409454 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00442321 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409505 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00946660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409446 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409475 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00103968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409462 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409508 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409302 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01138825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409432 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00357621 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00308467 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409291 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409433 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01138828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00705163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409423 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409466 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00334481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00588954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00308453 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409450 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409305 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409501 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00308465 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00308458 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409293 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409443 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409412 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409426 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409506 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409297 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409429 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00946646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409484 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409435 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00542403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409436 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00946652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00181030 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409425 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409300 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409512 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00705742 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409488 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409416 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00541333 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01136574 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00705165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00946635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409422 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409514 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20409299 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00308461 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10356301 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10356304 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10356303 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10356300 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10356302 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10356299 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100022125.mm.1 | NM_001033290.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100022125.mm.1 | NM_001033290.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100022125.mm.1 | TC0100002734.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100022127.mm.1 | NM_001033290.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100022127.mm.1 | NM_001033290.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100022127.mm.1 | TC0100002734.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100022126.mm.1 | NM_001033290.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100022126.mm.1 | NM_001033290.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100022126.mm.1 | TC0100002734.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100022124.mm.1 | NM_001033290.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100022124.mm.1 | NM_001033290.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100022124.mm.1 | TC0100002734.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1220868 | - | - |

Gpr55
Genomic Map