Assay Details
| Gene Symbol: Pram1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002842.2 | NP_001002842.2 | 5 - 6 | 1775 | 109 | |
| NR_028279.1 | 5 - 6 | 1771 | 109 | |||
| GenBank mRNA | AK169990.1 | - | 5 - 6 | 1767 | 109 | |
| AY665714.1 | - | 5 - 6 | 1691 | 109 | ||
| BC141100.1 | - | 5 - 6 | 1680 | 9055712 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999561 | AK169990.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999561 | AY665714.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999561 | NR_028279.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999561 | NM_001002842.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 753556 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 112955 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 436426 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 871606 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477031 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 85623 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P131237 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999561 | AK169990.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999561 | AY665714.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999561 | NR_028279.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999561 | NM_001002842.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149802 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149816 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149812 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149792 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149790 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149809 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149797 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149777 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P131237 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08763307 | - | - |
| Whole Mouse Genome G4122A | A_52_P306831 | BC141100.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P306831 | AK169990.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P306831 | AY665714.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P306831 | NR_028279.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P306831 | NM_001002842.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P15910 | AK169990.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P15910 | AY665714.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P15910 | NR_028279.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P15910 | NM_001002842.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110196 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P131237 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13220572 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13220558 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13220564 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13220560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149847 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149777 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13220570 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149824 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149812 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149854 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149791 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149779 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13220564 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13441354 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13234514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149830 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149809 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149847 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149831 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13106732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149837 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149775 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149833 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149805 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13352824 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149839 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149835 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149778 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13203162 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149774 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149784 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13234509 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149802 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149827 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13309191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149785 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13300367 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149819 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13220574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13441355 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149816 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13383587 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13498655 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149773 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13332187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149794 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149825 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13332181 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13332184 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149771 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149766 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13463556 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31149790 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10444025 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444019 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444024 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444017 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444026 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444027 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444018 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444022 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444023 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444021 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444020 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1444090_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10444016 | - | - |
| Mouse Genome 430 2.0 Array | 1444090_at | - | - |
| Mouse Genome 430 2.0 Array | 1444090_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 378460_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 378460_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700005409.mm.1 | NM_001002842.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005409.mm.1 | NM_001002842.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005409.mm.1 | NR_028279.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005409.mm.1 | NR_028279.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005409.mm.1 | TC1700002776.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700005402.mm.1 | NM_001002842.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005402.mm.1 | NM_001002842.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005402.mm.1 | NR_028279.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005402.mm.1 | NR_028279.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005402.mm.1 | TC1700002776.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700005405.mm.1 | NM_001002842.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005405.mm.1 | NM_001002842.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005405.mm.1 | NR_028279.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005405.mm.1 | NR_028279.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005405.mm.1 | TC1700002776.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700005406.mm.1 | NM_001002842.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005406.mm.1 | NM_001002842.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005406.mm.1 | NR_028279.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005406.mm.1 | NR_028279.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005406.mm.1 | TC1700002776.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700005407.mm.1 | NM_001002842.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005407.mm.1 | NM_001002842.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005407.mm.1 | NR_028279.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005407.mm.1 | NR_028279.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005407.mm.1 | TC1700002776.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700005408.mm.1 | NM_001002842.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005408.mm.1 | NM_001002842.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005408.mm.1 | NR_028279.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005408.mm.1 | NR_028279.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005408.mm.1 | TC1700002776.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700005400.mm.1 | NM_001002842.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005400.mm.1 | NM_001002842.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005400.mm.1 | NR_028279.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005400.mm.1 | NR_028279.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005400.mm.1 | TC1700002776.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700005404.mm.1 | NM_001002842.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005404.mm.1 | NM_001002842.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005404.mm.1 | NR_028279.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005404.mm.1 | NR_028279.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005404.mm.1 | TC1700002776.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1444090_PM_at | - | - |
| Mouse Expression Set 430 | 1444090_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444090_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444090_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2661567 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2661567 | - | - |

Pram1
Genomic Map