Assay Details
| Gene Symbol: Clpsl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034871.2 | NP_001030043.1 | 2 - 3 | 234 | 72 | |
| GenBank mRNA | AK136682.1 | - | 2 - 3 | 236 | 72 | |
| BC147116.1 | - | 2 - 3 | 310 | 9056160 | 72 | |
| BC147117.1 | - | 2 - 3 | 310 | 9056121 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087539 | NM_001034871.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087539 | AK136682.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087539 | BC147116.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087539 | BC147117.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149635 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 824614 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087539 | NM_001034871.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087539 | AK136682.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087539 | BC147116.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087539 | BC147117.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123037 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31123027 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08751057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13304055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13542038 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123038 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13542045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13308967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13542049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13308966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13575916 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13094437 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13542047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13575909 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31123060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13264182 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10443382 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443380 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443379 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10443378 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700004183.mm.1 | NM_001034871.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700004183.mm.1 | NM_001034871.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700004183.mm.1 | TC1700000482.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1214910 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2869966 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2869966 | - | - |

Clpsl2
Genomic Map