Assay Details
Gene Symbol: Krt40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039666.1 | NP_001034755.1 | 1 - 2 | 452 | 72 | |
GenBank mRNA | BC107359.1 | - | 1 - 2 | 452 | 72 | |
BC107360.2 | - | 1 - 2 | 450 | 72 | ||
BC119522.2 | - | 1 - 2 | 452 | 40055959 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
