Assay Details
| Gene Symbol: Gm15386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040027.2 | NP_001035116.1 | 1 - 2 | 113 | 90 | |
| GenBank mRNA | DQ012028.1 | - | 1 - 2 | 55 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069657 | NM_001040027.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069657 | DQ012028.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069657 | NM_001040027.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069657 | DQ012028.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20062613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20062598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20062614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20062611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20062606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20062607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20062604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20062612 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00085466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20062608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20062614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20062612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20062581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20062588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20062590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20062604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20062589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20062606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20062607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20062584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20062594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20062582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20062611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20062595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20062592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20062586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20062588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20062614 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20062581 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20062606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20062583 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20062590 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20062584 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20062586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20062605 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20062600 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20062595 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20062604 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20062611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20062607 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20062598 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20062594 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20062582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20062589 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20062612 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00085471 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20062592 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10353338 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10353339 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10353336 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100016512.mm.1 | NM_001040027.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100016512.mm.1 | NM_001040027.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100016512.mm.1 | NM_183124.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100016512.mm.1 | NM_183124.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100016512.mm.1 | TC0100002022.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100016510.mm.1 | NM_001040027.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100016510.mm.1 | NM_001040027.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100016510.mm.1 | NM_183124.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100016510.mm.1 | NM_183124.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100016510.mm.1 | TC0100002022.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100016509.mm.1 | NM_001040027.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100016509.mm.1 | NM_001040027.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100016509.mm.1 | TC0100002022.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2895546 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2895546 | - | - |

Gm15386
Genomic Map