Assay Details
Gene Symbol: Mrps33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010270.2 | NP_034400.1 | 2 - 3 | 310 | 81 | |
GenBank mRNA | AK019000.1 | - | 2 - 3 | 262 | 81 | |
AK144131.1 | - | 2 - 3 | 267 | 81 | ||
BC092135.1 | - | 1 - 2 | 214 | 5713592 | 81 | |
BC096397.1 | - | 1 - 2 | 257 | 3466818 | 81 | |
BY296250.1 | - | 2 - 3 | 305 | 81 | ||
Y17852.1 | - | 2 - 3 | 276 | 81 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24385098 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24385097 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24385095 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24385094 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P342896 | BC028645.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P342896 | BC087881.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P342896 | NM_001010930.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P342896 | BC028645.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P342896 | BC087881.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P342896 | NM_001010930.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2171715 | Y17852.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2171715 | BC028645.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2171715 | BC087881.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2171715 | BC092135.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2171715 | BC096397.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 152175 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 383280 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 609997 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 534340 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 650377 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 53034 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117848 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149791 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P342896 | BC028645.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P342896 | BC087881.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P342896 | NM_001010930.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P342896 | BC028645.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P342896 | BC087881.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P342896 | NM_001010930.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2171715 | Y17852.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2171715 | BC028645.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2171715 | BC087881.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2171715 | BC092135.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2171715 | BC096397.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P187093 | Y17852.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P187093 | BC028645.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P187093 | BC087881.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P187093 | BC092135.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P187093 | BC096397.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24385115 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24385092 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24385081 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24385102 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24385093 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24385100 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24385084 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24385086 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24385111 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24385109 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24385101 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24385082 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24385107 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24385090 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24385099 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24385110 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24385104 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24385089 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24385112 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24385105 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24385096 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24385085 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149791 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117848 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01310256 | - | - |
Whole Mouse Genome G4122A | A_51_P187093 | Y17852.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P187093 | BC028645.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P187093 | BC087881.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P187093 | BC092135.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P187093 | BC096397.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P579658 | BY296250.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P579658 | Y17852.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P579658 | BC092135.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P579658 | BC096397.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P579658 | NM_010270.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P342896 | BC028645.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P342896 | BC087881.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P342896 | NM_001010930.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P342896 | BC028645.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P342896 | BC087881.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P342896 | NM_001010930.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135701 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117848 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149791 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05744300 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05744312 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05744304 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05844190 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05744303 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05744313 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24385098 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24385095 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24385094 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24385097 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385072 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385074 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24384999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385126 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385004 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385081 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385002 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24384997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24384995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385062 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385082 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385102 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385067 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385040 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385115 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24385064 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385054 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05120890 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05748599 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385023 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385122 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385024 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385059 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05809529 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385092 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385034 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385105 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05644652 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385067 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385005 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385074 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385013 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385069 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385085 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385006 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385102 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05744300 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385111 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05744306 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05358612 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385078 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385004 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05263892 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385029 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05185975 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385045 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05558748 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385055 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385099 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05532770 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385032 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385112 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385056 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385017 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385086 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385031 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385080 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385052 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385026 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05748597 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385058 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385070 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385064 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385107 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385048 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385072 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385084 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385007 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385113 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385076 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385110 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385096 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385089 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385109 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05138357 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385015 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385012 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385050 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385062 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24385124 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10544246 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10563833 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10563831 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544245 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544247 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544244 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10563832 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10563830 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1427173_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10544243 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10563829 | - | - |
Mouse Genome 430 2.0 Array | 1427173_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1427173_a_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600016860.mm.1 | TC0600002133.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600016860.mm.1 | NM_010270.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600016860.mm.1 | NM_010270.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600016858.mm.1 | TC0600002133.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600016858.mm.1 | NM_001010930.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600016858.mm.1 | NM_001010930.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600016858.mm.1 | NM_010270.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600016858.mm.1 | NM_010270.2 | Refseq |
Mouse Expression Set 430 | 1427173_PM_a_at | - | - |
Mouse Expression Set 430 | 1427173_a_at | - | - |
Murine Genome U74v2 Set | 92826_PM_at | - | - |
Murine Genome U74v2 Set | 92826_at | - | - |
Mouse Genome 430A 2.0 Array | 1427173_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1427173_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427173_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427173_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 557165 | - | - |
ABI Mouse Genome Survey Microarray | 896827 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 5340722 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005340722 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1214937 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3163302 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_3163302 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1214937 | - | - |
mouse-6 v1.1 (Illumina) | GI_58331152-A | - | - |
mouse-6 v1.1 (Illumina) | scl014548.2_14-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5340722 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl014548.2_14-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5340722 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3163302 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1214937 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3163302 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1214937 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl014548.2_14-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | GI_58331152-A | - | - |
