Assay Details
| Gene Symbol: Ctdnep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026017.2 | NP_080293.1 | 6 - 7 | 904 | 85 | |
| GenBank mRNA | AK076150.1 | - | 6 - 7 | 916 | 85 | |
| AK164602.1 | - | 6 - 7 | 846 | 85 | ||
| BC018265.1 | - | 6 - 7 | 904 | 4486464 | 85 | |
More Information
Set Membership: |
|

Ctdnep1
Genomic Map