Assay Details
| Gene Symbol: Tas2r102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199153.2 | NP_954604.2 | 1 | 83 | 124 | |
| GenBank mRNA | BC117728.2 | - | 1 | 82 | 124 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124686 | NM_199153.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124686 | BC117728.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368656 | NM_001099305.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368656 | NM_001099306.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368656 | XM_001475692.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368656 | XM_903323.3 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368656 | XM_909963.3 | refseq_mrna_mouse_XM |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128623 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2124686 | NM_199153.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2124686 | BC117728.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P47461 | NM_199153.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P47461 | BC117728.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854577 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854573 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854574 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854569 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854570 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24854529 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128623 | - | - |
| Whole Mouse Genome G4122A | A_52_P47461 | NM_199153.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P47461 | BC117728.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128623 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05723885 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05487184 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05723886 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05418395 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05723874 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05723879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24854592 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854591 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854574 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05131630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854543 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05811800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05723883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854600 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854538 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05765131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05492646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854592 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05148104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05331021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854540 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854588 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24854550 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10542246 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10542245 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600012612.mm.1 | NM_199153.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012612.mm.1 | NM_199153.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012612.mm.1 | TC0600001562.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 625901 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5340647 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005340647 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2592404 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2592404 | - | - |
| mouse-6 v1.1 (Illumina) | scl0387339.1_310-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5340647 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0387339.1_310-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5340647 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2592404 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2592404 | - | - |

Tas2r102
Genomic Map