Assay Details
| Gene Symbol: Olfr685 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011857.1 | NP_001011857.1 | 1 | 693 | 93 | |
| GenBank mRNA | BC147196.1 | - | 1 | 782 | 9056252 | 93 |
| BC147197.1 | - | 1 | 782 | 9056258 | 93 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P467511 | BC147196.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P467511 | BC147197.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P467511 | NM_001011857.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P467511 | BC147196.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P467511 | BC147197.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P467511 | NM_001011857.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 254611 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P467511 | BC147196.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P467511 | BC147197.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P467511 | NM_001011857.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P467511 | BC147196.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P467511 | BC147197.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P467511 | NM_001011857.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P467511 | BC147196.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P467511 | BC147197.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P467511 | NM_001011857.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P467511 | BC147196.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P467511 | BC147197.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P467511 | NM_001011857.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391217 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391207 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391174 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391201 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25391170 | - | - |
| Whole Mouse Genome G4122A | A_51_P467511 | BC147196.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P467511 | BC147197.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P467511 | NM_001011857.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P467511 | BC147196.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P467511 | BC147197.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P467511 | NM_001011857.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06100028 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06100029 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06368221 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06368217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25391162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391201 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06648191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06413160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391161 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06433727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06321404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06413164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06433728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06368216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06080659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06648182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06413156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25391186 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10566431 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10566430 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700033058.mm.1 | NM_001011857.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700033058.mm.1 | NM_001011857.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700033058.mm.1 | TC0700003971.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700033059.mm.1 | TC0700003971.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700033057.mm.1 | TC0700003971.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3161860 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3161860 | - | - |
| mouse-6 v1.1 (Illumina) | GI_58801471-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3161860 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3161860 | - | - |

Olfr685
Genomic Map