Assay Details
| Gene Symbol: Zfp597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033159.2 | NP_001028331.2 | 2 - 3 | 437 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30506933 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30506931 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30506932 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30506929 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30506930 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031302 | BC057047.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031302 | BC108421.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031302 | NM_001033159.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 511883 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 897280 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 11272 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 698039 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 486974 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 108492 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140472 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031302 | BC057047.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031302 | BC108421.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031302 | NM_001033159.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506967 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506922 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506927 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506974 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506981 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30506923 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03120950 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08490953 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08490966 | - | - |
| Whole Mouse Genome G4122A | A_51_P149842 | AK016727.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P149842 | BC057047.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P149842 | BC108421.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P149842 | NM_001033159.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08490966 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30506930 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30506932 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30506931 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30506933 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30506929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30506946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506898 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506909 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12553190 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506957 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506929 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12620476 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13008968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506927 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12553184 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12476849 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506902 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506919 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506923 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506963 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12553187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506951 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506897 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506962 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13036484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506906 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12983433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12620475 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506920 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506945 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30506959 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10437261 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437260 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437262 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437259 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437258 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430184_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435556_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460589_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1443102_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10437257 | - | - |
| Mouse Genome 430 2.0 Array | 1443102_at | - | - |
| Mouse Genome 430 2.0 Array | 1435556_at | - | - |
| Mouse Genome 430 2.0 Array | 1443102_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430184_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1460589_at | - | - |
| Mouse Genome 430 2.0 Array | 1460589_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1435556_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430184_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71063_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71063_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600008723.mm.1 | NM_001033159.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008723.mm.1 | NM_001033159.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008723.mm.1 | TC1600001107.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600008718.mm.1 | NM_001033159.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008718.mm.1 | NM_001033159.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008718.mm.1 | TC1600001107.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600008727.mm.1 | NM_001033159.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008727.mm.1 | NM_001033159.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008727.mm.1 | TC1600001107.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600008717.mm.1 | NM_001033159.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008717.mm.1 | NM_001033159.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008717.mm.1 | TC1600001107.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600008721.mm.1 | NM_001033159.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008721.mm.1 | NM_001033159.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008721.mm.1 | TC1600001107.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600008722.mm.1 | NM_001033159.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008722.mm.1 | NM_001033159.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008722.mm.1 | TC1600001107.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600008725.mm.1 | NM_001033159.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008725.mm.1 | NM_001033159.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008725.mm.1 | TC1600001107.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600008719.mm.1 | NM_001033159.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008719.mm.1 | NM_001033159.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008719.mm.1 | TC1600001107.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600008720.mm.1 | NM_001033159.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008720.mm.1 | NM_001033159.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008720.mm.1 | TC1600001107.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600008724.mm.1 | NM_001033159.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008724.mm.1 | NM_001033159.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008724.mm.1 | TC1600001107.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600008716.mm.1 | NM_001033159.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008716.mm.1 | NM_001033159.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008716.mm.1 | TC1600001107.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1435556_at | - | - |
| Mouse Expression Set 430 | 1460589_at | - | - |
| Mouse Expression Set 430 | 1430184_at | - | - |
| Mouse Expression Set 430 | 1430184_PM_at | - | - |
| Mouse Expression Set 430 | 1435556_PM_at | - | - |
| Mouse Expression Set 430 | 1443102_PM_at | - | - |
| Mouse Expression Set 430 | 1460589_PM_at | - | - |
| Mouse Expression Set 430 | 1443102_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443102_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443102_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435556_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1460589_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430184_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430184_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435556_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1460589_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 784560 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3160688 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3160688 | - | - |
| mouse-6 v1.1 (Illumina) | GI_84370271-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3160688 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3160688 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | GI_84370271-S | - | - |

Zfp597
Genomic Map