Assay Details
| Gene Symbol: Nodal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013611.4 | NP_038639.2 | 1 - 2 | 523 | 109 | |
| GenBank mRNA | AK049290.1 | - | 1 - 2 | 207 | 109 | |
| AK082823.1 | - | 1 - 2 | 207 | 109 | ||
| BC128018.1 | - | 1 - 2 | 200 | 40136148 | 109 | |
| X70514.1 | - | 1 - 2 | 522 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nodal
Genomic Map