Assay Details
| Gene Symbol: Gm732 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033252.2 | NP_001028424.1 | 2 | 1590 | 93 | |
| GenBank mRNA | AK132985.1 | - | 2 | 1592 | 93 | |
| BC147603.1 | - | 2 | 1529 | 9056593 | 93 | |
| BC147609.1 | - | 2 | 1529 | 9056599 | 93 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P979001 | NM_001033252.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P979001 | AK132985.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P979001 | BC147603.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P979001 | BC147609.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P979001 | NM_001033252.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P979001 | AK132985.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P979001 | BC147603.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P979001 | BC147609.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 771855 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 306624 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 155875 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P979001 | NM_001033252.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P979001 | AK132985.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P979001 | BC147603.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P979001 | BC147609.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P979001 | NM_001033252.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P979001 | AK132985.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P979001 | BC147603.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P979001 | BC147609.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03734893 | - | - |
| Whole Mouse Genome G4122A | A_52_P426316 | NM_001033252.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P426316 | AK132985.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P426316 | BC147603.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P426316 | BC147609.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P979001 | NM_001033252.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P979001 | AK132985.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P979001 | BC147603.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P979001 | BC147609.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P979001 | NM_001033252.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P979001 | AK132985.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P979001 | BC147603.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P979001 | BC147609.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03734893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32590232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590233 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15142870 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14744386 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590214 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590245 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14609355 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590200 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14958639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15142864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15163756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590246 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590244 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590203 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590234 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590211 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590239 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15142858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590216 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590192 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15120757 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590184 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590212 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32590202 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10606387 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606388 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1445435_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10606386 | - | - |
| Mouse Genome 430 2.0 Array | 1445435_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1445435_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 213450_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 213450_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00018113.mm.1 | NM_001033252.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00018113.mm.1 | NM_001033252.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00018113.mm.1 | TC0X00002848.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00018112.mm.1 | NM_001033252.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00018112.mm.1 | NM_001033252.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00018112.mm.1 | TC0X00002848.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00018115.mm.1 | NM_001033252.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00018115.mm.1 | NM_001033252.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00018115.mm.1 | TC0X00002848.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00018111.mm.1 | NM_001033252.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00018111.mm.1 | NM_001033252.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00018111.mm.1 | TC0X00002848.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1445435_PM_at | - | - |
| Mouse Expression Set 430 | 1445435_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445435_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445435_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 305924 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1229474 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3022025 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3094365 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3094365 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3022025 | - | - |

Gm732
Genomic Map